Ethyl-1-benzylpyrrole-3-carboxylate ≥95.0%

Supplier: TCI America

B2284-1G B2284-5G
TCB2284-1GEA 153.26 CAD
TCB2284-1G TCB2284-5G
Ethyl-1-benzylpyrrole-3-carboxylate ≥95.0%
Ethyl-1-benzylpyrrole-3-carboxylate

CAS Number: 128259-47-2
MDL Number: MFCD06797056
Molecular Formula: C14H15NO2
Molecular Weight: 229.28
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.11
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1-Benzylpyrrole-3-carboxylic Acid Ethyl Ester

Formula: C₁₄H₁₅NO₂
Density: 1.11 (20 °C)
MDL Number: MFCD06797056
CAS Number: 128259-47-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c7a3687ecd4d827f52052850b7484f1c7c75482&vid=6c7a3687ecd4d827f52052850b7484f1c7c75482&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR