2,6-Bis(4-fluorobenzylidene)cyclohexanone ≥96.0% (by HPLC)

Supplier: TCI America

Synonyms: 2,6-Bis(4-fluorobenzal)cyclohexanone

B2282-25G B2282-5G
TCB2282-25GEA 365.3 CAD
TCB2282-25G TCB2282-5G
2,6-Bis(4-fluorobenzylidene)cyclohexanone ≥96.0% (by HPLC)
2,6-Bis(4-fluorobenzylidene)cyclohexanone

CAS Number: 62085-74-9
MDL Number: MFCD00112608
Molecular Formula: C20H16F2O
Molecular Weight: 310.34
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,6-Bis(4-fluorobenzal)cyclohexanone

Formula: C₂₀H₁₆F₂O
MW: 310.34 g/mol
MDL Number: MFCD00112608
CAS Number: 62085-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c79d3caae0ca1e76145067b22c6d759c98d7798&vid=2c79d3caae0ca1e76145067b22c6d759c98d7798&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR