1,1'-[Biphenyl-4,4'-diylbis(methylene)]bis(4,4'-bipyridinium)dibromide ≥96.0%

Supplier: TCI America

B2279-100MG B2279-1G
TCB2279-100MGEA 67.78 CAD
TCB2279-100MG TCB2279-1G
1,1'-[Biphenyl-4,4'-diylbis(methylene)]bis(4,4'-bipyridinium)dibromide ≥96.0%
1,1'-[Biphenyl-4,4'-diylbis(methylene)]bis(4,4'-bipyridinium)dibromide

MDL Number: MFCD06797054
Molecular Formula: C34H28Br2N4
Molecular Weight: 652.43
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal

Formula: C₃₄H₂₈Br₂N₄
MW: 652.43 g/mol
MDL Number: MFCD06797054
CAS Number: 2232875-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51cc32be97f0abb582e1e176c560dd07c26e072e&vid=51cc32be97f0abb582e1e176c560dd07c26e072e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR