2-Benzyloxy-1,3-propanediol ≥97.0%
Supplier: TCI America
Synonyms:
2-(Benzyloxy)propane-1,3-diol, 2-O-Benzylglycerol, Glycerol 2-Benzyl Ether
B2271-1G
B2271-5G
TCB2271-1GEA
344.44
CAD
TCB2271-1G
TCB2271-5G
2-Benzyloxy-1,3-propanediol ≥97.0%
2-Benzyloxy-1,3-propanediol
CAS Number: 14690-00-7
MDL Number: MFCD00075493
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 42
Flash Point (°C): 110
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2-O-Benzylglycerol, Glycerol 2-Benzyl Ether
Formula:
C₁₀H₁₄O₃ MW: 182.22 g/mol Melting Pt: 42 °C Flash Pt: 110 °C |
MDL Number:
MFCD00075493 CAS Number: 14690-00-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...