4,4'-Bis[di(3,5-xylyl)amino]-4''-phenyltriphenylamine ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
N-(4-Biphenylyl)-N,N-bis[4-[di(3,5-xylyl)amino]phenyl]amine
B2269-100MG
B2269-1G
TCB2269-100MGEA
497.54
CAD
TCB2269-100MG
TCB2269-1G
4,4'-Bis[di(3,5-xylyl)amino]-4''-phenyltriphenylamine ≥98.0% (by HPLC)
4,4'-Bis[di(3,5-xylyl)amino]-4''-phenyltriphenylamine
CAS Number: 249609-49-2
MDL Number: MFCD03844780
Molecular Formula: C56H53N3
Molecular Weight: 768.06
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials
Other Product Names / Synonyms: N-(4-Biphenylyl)-N,N-bis[4-[di(3,5-xylyl)amino]phenyl]amine
Formula:
C₅₆H₅₃N₃ MW: 768.06 g/mol |
MDL Number:
MFCD03844780 CAS Number: 249609-49-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...