10-Bromodecanoic acid ≥97.0%
Supplier: TCI America
B2264-25G
B2264-5G
TCB2264-25GEA
213.3
CAD
TCB2264-25G
TCB2264-5G
10-Bromodecanoic acid ≥97.0%
10-Bromodecanoic acid
CAS Number: 50530-12-6
MDL Number: MFCD00014388
Molecular Formula: C10H19BrO2
Molecular Weight: 251.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 42
Flash Point (°C): 110
Formula:
C₁₀H₁₉BrO₂ Melting Pt: 42 °C Flash Pt: 110 °C |
MDL Number:
MFCD00014388 CAS Number: 50530-12-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...