Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B2253-5G B2253-25G
TCB2253-5GEA 67.78 CAD
TCB2253-5G TCB2253-25G
Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine ≥98.0% (by HPLC, titration analysis)
Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine

CAS Number: 2480-93-5
MDL Number: MFCD00038259
Molecular Formula: C18H26N2O6
Molecular Weight: 366.41
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 94
Specific rotation [a]20/D: -3 deg (C=1, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Boc-Ndelta-Cbz-L-ornithine, Boc-Orn(Z)-OH

Formula: C₁₈H₂₆N₂O₆
Melting Pt: 94 °C
MDL Number: MFCD00038259
CAS Number: 2480-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=340ff829f39dafda3c31ba07dccdc9227d03ae5d&vid=340ff829f39dafda3c31ba07dccdc9227d03ae5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR