3,5-Bis(3,5-dimethoxybenzyloxy)benzyl bromide ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
B2247-1G
B2247-5G
TCB2247-1GEA
317.42
CAD
TCB2247-1G
TCB2247-5G
3,5-Bis(3,5-dimethoxybenzyloxy)benzyl bromide ≥97.0% (by HPLC, titration analysis)
3,5-Bis(3,5-dimethoxybenzyloxy)benzyl bromide
CAS Number: 176650-93-4
MDL Number: MFCD04038410
Molecular Formula: C25H27BrO6
Molecular Weight: 503.39
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C
Formula:
C₂₅H₂₇BrO₆ MW: 503.39 g/mol |
MDL Number:
MFCD04038410 CAS Number: 176650-93-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...