3,5-Bis(3,5-dimethoxybenzyloxy)benzyl alcohol ≥94.0%

Supplier: TCI America

B2246-5G B2246-1G
TCB2246-5GEA 328.96 CAD
TCB2246-5G TCB2246-1G
3,5-Bis(3,5-dimethoxybenzyloxy)benzyl alcohol ≥94.0%
3,5-Bis(3,5-dimethoxybenzyloxy)benzyl alcohol

CAS Number: 176650-92-3
MDL Number: MFCD03844779
Molecular Formula: C25H28O7
Molecular Weight: 440.49
Purity/Analysis Method: >94.0% (GC)
Form: Crystal

Formula: C₂₅H₂₈O₇
MDL Number: MFCD03844779
CAS Number: 176650-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=175a0e60c853adc4642123b7b6862ed11f616de9&vid=175a0e60c853adc4642123b7b6862ed11f616de9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR