3'-Bromo-5'-chloro-2'-hydroxyacetophenone ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B2236-5G B2236-25G
TCB2236-5GEA 118.97 CAD
TCB2236-5G TCB2236-25G
3'-Bromo-5'-chloro-2'-hydroxyacetophenone ≥97.0% (by GC, titration analysis)
3'-Bromo-5'-chloro-2'-hydroxyacetophenone

CAS Number: 59443-15-1
MDL Number: MFCD00857068
Molecular Formula: C8H6BrClO2
Molecular Weight: 249.49
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 100


Other Product Names / Synonyms: 2-Acetyl-6-bromo-4-chlorophenol

Formula: C₈H₆BrClO₂
Melting Pt: 100 °C
MDL Number: MFCD00857068
CAS Number: 59443-15-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0cdba1053e1c63040a715528688672cf872f8bf&vid=c0cdba1053e1c63040a715528688672cf872f8bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR