3-(Boc-amino)propylbromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-(tert-Butoxycarbonylamino)propyl bromide, 3-(Boc-amino)propyl bromide, tert-Butyl N-(3-Bromopropyl)carbamate, N-(3-Bromopropyl)carbamic acid tert-Butyl ester

B2234-1G B2234-5G
TCB2234-1GEA 292.3 CAD
TCB2234-1G TCB2234-5G
3-(Boc-amino)propylbromide ≥98.0% (by titrimetric analysis)
3-(Boc-amino)propylbromide

CAS Number: 83948-53-2
MDL Number: MFCD02683429
Molecular Formula: C8H16BrNO2
Molecular Weight: 238.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 41
Storage Temperature: <0°C


Other Product Names / Synonyms: N-(3-Bromopropyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(3-Bromopropyl)carbamate, 3-(Boc-amino)propyl Bromide

Formula: C8H16BrNO2
MW: 238.1221 g/mol
Melting Pt: 41 °C
MDL Number: MFCD02683429
CAS Number: 83948-53-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a124580ad539946ab5ba6e2dba2bf12927da10f9&vid=a124580ad539946ab5ba6e2dba2bf12927da10f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR