(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid ≥98.0%

Supplier: TCI America

Synonyms: Boc-Phe(4-NO2)-OH, Boc-Phe(4-NO₂)-OH, N-Boc-4-nitro-L-phenylalanine, Boc-p-nitro-Phe-OH

B2232-1G B2232-5G
TCB2232-1GEA 36.34 CAD
TCB2232-1G TCB2232-5G
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid ≥98.0%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid

CAS Number: 33305-77-0
MDL Number: MFCD00038128
Molecular Formula: C14H18N2O6
Molecular Weight: 310.31
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 110
Specific rotation [a]20/D: 8 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-4-nitro-L-phenylalanine, Boc-Phe(4-NO2)-OH

Formula: C₁₄H₁₈N₂O₆
MW: 310.3 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00038128
CAS Number: 33305-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37b7e49a4c198660521072340fcfe2da85d0bdaa&vid=37b7e49a4c198660521072340fcfe2da85d0bdaa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR