2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥95.0%

Supplier: TCI America

Synonyms: (S,S)-Ph-pybox, (-)-2,6-Bis[(4S)-4-phenyl-2-oxazolin-2-yl]pyridine

B2220-250MG B2220-1G
TCB2220-250MGEA 218.04 CAD
TCB2220-250MG TCB2220-1G
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥95.0%
2,6-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine

CAS Number: 174500-20-0
MDL Number: MFCD01863586
Molecular Formula: C23H19N3O2
Molecular Weight: 369.42
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 164
Specific rotation [a]20/D: -175 deg (C=1, CH2Cl2)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (S,S)-2,2'-(2,6-Pyridinediyl)bis(4-phenyl-2-oxazoline)

Formula: C₂₃H₁₉N₃O₂
Melting Pt: 164 °C
MDL Number: MFCD01863586
CAS Number: 174500-20-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e501756a3430e33b9659a959b4908e1ec131590&vid=7e501756a3430e33b9659a959b4908e1ec131590&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR