2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B2219-1G B2219-250MG
TCB2219-1GEA 278.08 CAD
TCB2219-1G TCB2219-250MG
2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98.0% (by HPLC, titration analysis)
2,6-Bis((R)-4-phenyl-4,5-dihydrooxazol-2-yl)pyridine

CAS Number: 128249-70-7
MDL Number: MFCD01863585
Molecular Formula: C23H19N3O2
Molecular Weight: 369.42
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 173
Specific rotation [a]20/D: 190 deg (C=1, CH2Cl2)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (R,R)-2,2'-(2,6-Pyridinediyl)bis(4-phenyl-2-oxazoline)

Formula: C₂₃H₁₉N₃O₂
Melting Pt: 173 °C
MDL Number: MFCD01863585
CAS Number: 128249-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=096fd388e98d39f4eedac9b3686b0091b51774fc&vid=096fd388e98d39f4eedac9b3686b0091b51774fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR