(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98.0%

Supplier: TCI America

Synonyms: (-)-2,6-Bis[(4S)-4-(i-propyl)-2-oxazolin-2-yl]pyridine, (S,S)-2,2'-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline), 2,6-Bis((S)-4-isopropyl-4,5-dihydrooxazol-2-yl)pyridine, (S)-(i-Pr)-pybox

B2218-500MG B2218-5G
TCB2218-500MGEA 162.74 CAD
TCB2218-500MG TCB2218-5G
(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98.0%
(S,S)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine

CAS Number: 118949-61-4
MDL Number: MFCD00191815
Molecular Formula: C17H23N3O2
Molecular Weight: 301.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 152
Specific rotation [a]20/D: -120 deg (C=1, CH2Cl2)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (S,S)-2,2'-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline)

Formula: C₁₇H₂₃N₃O₂
MW: 301.39 g/mol
Melting Pt: 152 °C
MDL Number: MFCD00191815
CAS Number: 118949-61-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR