5-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B2214-25G
B2214-5G
TCB2214-25GEA
1036.48
CAD
TCB2214-25G
TCB2214-5G
5-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)
5-Bromo-2-hydroxybenzonitrile
CAS Number: 40530-18-5
MDL Number: MFCD00143428
Molecular Formula: C7H4BrNO
Molecular Weight: 198.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 162
Other Product Names / Synonyms: 4-Bromo-2-cyanophenol, 2-Hydroxy-5-bromobenzonitrile
Formula:
C₇H₄BrNO Melting Pt: 162 °C |
MDL Number:
MFCD00143428 CAS Number: 40530-18-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...