Triethyl acetylcitrate ≥97.0% for synthesis
Supplier: TCI America
Synonyms:
O-Acetylcitric acid triethyl ester, Triethyl O-Acetylcitrate, Acetyltriethylcitrate, Triethyl 2-acetoxypropane-1,2,3-tricarboxylate
CAS Number: 77-89-4
MDL Number: MFCD00049378
Molecular Formula: C14H22O8
Molecular Weight: 318.32
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 229
Flash Point (°C): 188
Specific Gravity (20/20): 1.14
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: O-Acetylcitric Acid Triethyl Ester
Formula:
C₁₄H₂₂O₈ Boiling Pt: 229 °C Density: 1.14 (20 °C) Flash Pt: 188 °C |
MDL Number:
MFCD00049378 CAS Number: 77-89-4 EINECS: 201-066-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...