Boc-D-Arg-OH·HCl·H2O ≥98.0% (by total nitrogen basis)

Supplier: TCI America

B2204-1G B2204-5G
TCB2204-1GEA 62.88 CAD
TCB2204-1G TCB2204-5G
Boc-D-Arg-OH·HCl·H2O ≥98.0% (by total nitrogen basis)
Boc-D-Arg-OH·HCl·H2O

CAS Number: 114622-81-0
MDL Number: MFCD00065554
Molecular Formula: C11H22N4O4
Molecular Weight: 310.78
Purity/Analysis Method: >98.0% (N)
Form: Crystal


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Boc-D-arginine Hydrochloride, Boc-D-Arg-OH.HCl

Formula: C₁₁H₂₂N₄O₄·HCl·H₂O
MDL Number: MFCD00065554
CAS Number: 204070-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=375f0c9718079e1c87e14bce54eba3f004d94560&vid=375f0c9718079e1c87e14bce54eba3f004d94560&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR