4'-Bromobenzo-15-crown-5 (4'-bromobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥93.0%

Supplier: TCI America

Synonyms: 4'-Bromobenzo-15-crown 5-Ether, 2,3-(4-Bromobenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene, 15-Bromo-2,3,5,6,8,9,11,12-octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine

B2189-1G B2189-5G
TCB2189-1GEA 398.16 CAD
TCB2189-1G TCB2189-5G
4'-Bromobenzo-15-crown-5 (4'-bromobenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥93.0%
4'-Bromobenzo-15-crown-5 (4'-Bromobenzo-1,4,7,10,13-pentaoxacyclopentadecane)

CAS Number: 60835-72-5
MDL Number: MFCD00143270
Molecular Formula: C14H19BrO5
Molecular Weight: 347.21
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 70


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-(4-Bromobenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene

Formula: C₁₄H₁₉BrO₅
MW: 347.2000 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00143270
CAS Number: 60835-72-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9aa8c8da44845dc356cd73ed66f294c6b7b1f3e2&vid=9aa8c8da44845dc356cd73ed66f294c6b7b1f3e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR