6-Bromo-3-formylchromone ≥97.0%

Supplier: TCI America

B2183-25G B2183-5G
TCB2183-25GEA 244.58 CAD
TCB2183-25G TCB2183-5G
6-Bromo-3-formylchromone ≥97.0%
6-Bromo-3-formylchromone

CAS Number: 52817-12-6
MDL Number: MFCD00191849
Molecular Formula: C10H5BrO3
Molecular Weight: 253.05
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 191


Other Product Names / Synonyms: 6-Bromo-4-oxo-4H-1-benzopyran-3-carboxaldehyde, 6-Bromochromone-3-carboxaldehyde

Formula: C₁₀H₅BrO₃
Melting Pt: 191 °C
MDL Number: MFCD00191849
CAS Number: 52817-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33986b9725eceff809f055f34eb3f5325778c133&vid=33986b9725eceff809f055f34eb3f5325778c133&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR