4'-Bromobenzo-18-crown-6 (4'-bromobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥95.0%

Supplier: TCI America

Synonyms: 18-Bromo-2,3,5,6,8,9,11,12,14,15-decahydrobenzo[b][1,4,7,10,13,16]hexaoxacyclooctadecine, 4'-Bromobenzo-18-crown 6-Ether, 2,3-(4-Bromobenzo)-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene

B2181-5G B2181-1G
TCB2181-5GEA 603.56 CAD
TCB2181-5G TCB2181-1G
4'-Bromobenzo-18-crown-6 (4'-bromobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥95.0%
4'-Bromobenzo-18-crown-6 (4'-Bromobenzo-1,4,7,10,13,16-hexoxacyclooctadecane)

CAS Number: 75460-28-5
MDL Number: MFCD00143200
Molecular Formula: C16H23BrO6
Molecular Weight: 391.26
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 80


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-(4-Bromobenzo)-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene

Formula: C₁₆H₂₃BrO₆
MW: 391.0000 g/mol
Melting Pt: 80 °C
MDL Number: MFCD00143200
CAS Number: 75460-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99bb3c4f95002418f1d171b2eee9d7e87ee50798&vid=99bb3c4f95002418f1d171b2eee9d7e87ee50798&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR