DL-Arginine hydrochloride ≥97.0%

Supplier: TCI America

Synonyms: DL-Arginine xhydrochloride, DL-Arginine Hydrochloride, DL-(+/-)-Arginine hydrochloride, DL-(±)-Arginine Hydrochloride, H-DL-Arg-OH.HCl, H-DL-Arg-OH·HCl, H-DL-(±)-Arg-OH·HCl, DL-2-Amino-5-guanidinopentanoic acid hydrochloride monohydrate

A0527-1G A0527-25G A0527-5G
TCA0527-001GEA 66.36 CAD
TCA0527-001G TCA0527-025G TCA0527-5G
DL-Arginine hydrochloride ≥97.0%
DL-Arginine hydrochloride

CAS Number: 32042-43-6
MDL Number: MFCD00064549
Molecular Formula: C6H14N4O2
Molecular Weight: 210.66
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Arg-OH.HCl

Formula: C₆H₁₄N₄O₂.HCl
MDL Number: MFCD00064549
CAS Number: 32042-43-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8aabd4a26c7f14b5a6027a4e4c931dc6100c010&vid=f8aabd4a26c7f14b5a6027a4e4c931dc6100c010&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR