Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt ≥98.0%

Supplier: TCI America

Synonyms: Boc-Lys(Boc)-OH.DCHA, Boc-Lys(Boc)-OH·DCHA, Nα,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, Nɑ,N-ε-Di-Boc-L-lysine Dicyclohexylammonium Salt

B2178-25G B2178-5G
TCB2178-25GEA 91.64 CAD
TCB2178-25G TCB2178-5G
Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt ≥98.0%
Nalpha,N-epsilon-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt

CAS Number: 15098-69-8
MDL Number: MFCD00038892
Molecular Formula: C28H53N3O6
Molecular Weight: 527.75
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 141
Specific rotation [a]20/D: -6 deg (C=3, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha,Nepsilon-Di-Boc-L-lysine Dicyclohexylammonium Salt, Boc-Lys(Boc)-OH.DCHA

Formula: C₂₈H₅₃N₃O₆
MW: 527.75 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00038892
CAS Number: 15098-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cfd631acc1b81bdea0d02d7bbdd76b88221362f&vid=9cfd631acc1b81bdea0d02d7bbdd76b88221362f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR