(R)-1-Boc-2-Azetidinemethanol ≥98.0%
Supplier: TCI America
B2174-1G
B2174-100MG
TCB2174-1GEA
3105.81
CAD
TCB2174-1G
TCB2174-100MG
(R)-1-Boc-2-Azetidinemethanol ≥98.0%
(R)-1-Boc-2-Azetidinemethanol
CAS Number: 161511-90-6
MDL Number: MFCD03790937
Molecular Formula: C9H17NO3
Molecular Weight: 187.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific rotation [a]20/D: 75 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1-Boc-2-azetidinemethanol
Formula:
C₉H₁₇NO₃ |
MDL Number:
MFCD03790937 CAS Number: 161511-90-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...