4-Bromobenzene-1,2-diol ≥98.0%
Supplier: TCI America
B2173-1G
B2173-5G
TCB2173-1GEA
74.26
CAD
TCB2173-1G
TCB2173-5G
4-Bromobenzene-1,2-diol ≥98.0%
4-Bromobenzene-1,2-diol
CAS Number: 17345-77-6
MDL Number: MFCD00869769
Molecular Formula: C6H5BrO2
Molecular Weight: 189.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 87
Other Product Names / Synonyms: 4-Bromo-1,2-dihydroxybenzene
Formula:
C₆H₅BrO₂ Melting Pt: 87 °C |
MDL Number:
MFCD00869769 CAS Number: 17345-77-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...