N-Bromosaccharin ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2152-25G B2152-5G
TCB2152-25GEA 1075.98 CAD
TCB2152-25G TCB2152-5G
N-Bromosaccharin ≥98.0% (by titrimetric analysis)
N-Bromosaccharin

CAS Number: 35812-01-2
MDL Number: MFCD03844772
Molecular Formula: C7H4BrNO3S
Molecular Weight: 262.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: 2-Bromo-1,2-benzisothiazol-3(2H)-one 1,1-Dioxide

Formula: C₇H₄BrNO₃S
MDL Number: MFCD03844772
CAS Number: 35812-01-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af55ab4ce523bb914cac460d3631e77bdc9a8791&vid=af55ab4ce523bb914cac460d3631e77bdc9a8791&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR