(S)-(-)-3-Benzyloxy-1,2-propanediol ≥98.0%
Supplier: TCI America
B2142-100MG
B2142-1G
TCB2142-100MGEA
51.41
CAD
TCB2142-100MG
TCB2142-1G
(S)-(-)-3-Benzyloxy-1,2-propanediol ≥98.0%
(S)-(-)-3-Benzyloxy-1,2-propanediol
CAS Number: 17325-85-8
MDL Number: MFCD00077914
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 24
Specific rotation [a]20/D: -5.5 deg (C=20, CHCl3)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-Glycerol alpha-Benzyl Ether
Formula:
C₁₀H₁₄O₃ Flash Pt: 110 °C |
MDL Number:
MFCD00077914 CAS Number: 17325-85-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...