(R)-(+)-3-Benzyloxy-1,2-propanediol ≥98.0%

Supplier: TCI America

B2141-1G B2141-100MG
TCB2141-1GEA 184.86 CAD
TCB2141-1G TCB2141-100MG
(R)-(+)-3-Benzyloxy-1,2-propanediol ≥98.0%
(R)-(+)-3-Benzyloxy-1,2-propanediol

CAS Number: 56552-80-8
MDL Number: MFCD00067260
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific rotation [a]20/D: 5.5 deg (C=20, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-Glycerol alpha-Benzyl Ether

Formula: C₁₀H₁₄O₃
Flash Pt: 110 °C
MDL Number: MFCD00067260
CAS Number: 56552-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f3d04defc1e865056d8f546d11fe13905d47791&vid=3f3d04defc1e865056d8f546d11fe13905d47791&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR