Methyl (S)-(-)-3-bromoisobutyrate ≥97.0%

Supplier: TCI America

Synonyms: (S)-(-)-3-Bromoisobutyric Acid Methyl Ester, Methyl (S)-(-)-3-Bromo-2-methylpropionate, (S)-(-)-3-Bromo-2-methylpropionic Acid Methyl Ester, Methyl (S)-3-bromo-2-methylpropanoate

B2140-1G B2140-5G
TCB2140-1GEA 248.06 CAD
TCB2140-1G TCB2140-5G
Methyl (S)-(-)-3-bromoisobutyrate ≥97.0%
Methyl (S)-(-)-3-bromoisobutyrate

CAS Number: 98190-85-3
MDL Number: MFCD00064487
Molecular Formula: C5H9BrO2
Molecular Weight: 181.03
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 73
Specific Gravity (20/20): 1.43
Specific rotation [a]20/D: -16 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-3-Bromo-2-methylpropionic Acid Methyl Ester, (S)-(-)-3-Bromoisobutyric Acid Methyl Ester, Methyl (S)-(-)-3-Bromo-2-methylpropionate

Formula: C₅H₉BrO₂
Density: 1.43 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00064487
CAS Number: 98190-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f578963a0dc48fa3191b8fd6f6947d54a79ccb7c&vid=f578963a0dc48fa3191b8fd6f6947d54a79ccb7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR