Tetrasodium-N,N-bis(carboxymethyl)-L-glutamate ∼40% in water
Supplier: TCI America
B2135-25G
B2135-500G
TCB2135-025GEA
56.25
CAD
TCB2135-025G
TCB2135-500G
Tetrasodium-N,N-bis(carboxymethyl)-L-glutamate ∼40% in water
Tetrasodium-N,N-bis(carboxymethyl)-L-glutamate
CAS Number: 51981-21-6
MDL Number: MFCD01862262
Molecular Formula: C9H13NO8
Molecular Weight: 351.13
Form: Clear Liquid
Color: Slightly Pale Yellow
Specific rotation [a]20/D: 10 deg (neat)
Other Product Names / Synonyms: N,N-Bis(carboxymethyl)-L-glutamic Acid Tetrasodium Salt
Formula:
C₉H₁₃NO₈ |
MDL Number:
MFCD01862262 CAS Number: 51981-21-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...