1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1,1-Binaphthyl-2,2-diyl hydrogenphosphate
B2130-5G
B2130-25G
TCB2130-005GEA
133.35
CAD
TCB2130-005G
TCB2130-025G
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate ≥98.0% (by HPLC, titration analysis)
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
CAS Number: 35193-63-6
MDL Number: MFCD00010045
Molecular Formula: C20H13O4P
Molecular Weight: 348.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: (+/-)-2,2'-Dihydroxy-1,1'-binaphthyl Hydrogenphosphate
Formula:
C₂₀H₁₃O₄P MW: 348.29 g/mol |
MDL Number:
MFCD00010045 CAS Number: 35193-63-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...