2-(4-Bromobutyl)-1,3-dioxolane ≥95.0%
Supplier: TCI America
B2128-5G
B2128-25G
TCB2128-005GEA
352.02
CAD
TCB2128-005G
TCB2128-25G
2-(4-Bromobutyl)-1,3-dioxolane ≥95.0%
2-(4-Bromobutyl)-1,3-dioxolane
CAS Number: 87227-41-6
MDL Number: MFCD02093454
Molecular Formula: C7H13BrO2
Molecular Weight: 209.08
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.38
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 5-Bromovaleraldehyde Ethylene Acetal
Formula:
C₇H₁₃BrO₂ Density: 1.38 (20 °C) |
MDL Number:
MFCD02093454 CAS Number: 87227-41-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...