4-[(tert-Butoxycarbonylamino)methyl]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-(Boc-aminomethyl)pyridine

B2126-25G B2126-5G
TCB2126-25GEA 665.18 CAD
TCB2126-25G TCB2126-005G
4-[(tert-Butoxycarbonylamino)methyl]pyridine ≥98.0% (by GC, titration analysis)
4-[(tert-Butoxycarbonylamino)methyl]pyridine

CAS Number: 111080-65-0
MDL Number: MFCD01321573
Molecular Formula: C11H16N2O2
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 91


Other Product Names / Synonyms: 4-(Boc-aminomethyl)pyridine

Formula: C₁₁H₁₆N₂O₂
MW: 208.26 g/mol
Melting Pt: 91 °C
MDL Number: MFCD01321573
CAS Number: 111080-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21b9ca25967366d7eca454d93dd403daf167b95b&vid=41e7e1a9c1acbda29ddb3df011a6edebca640fdb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR