4-[(tert-Butoxycarbonylamino)methyl]pyridine-2-carboxylic acid ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-(Boc-aminomethyl)pyridine-2-carboxylic Acid

B2124-1G B2124-100MG
TCB2124-1GEA 1191.79 CAD
TCB2124-1G TCB2124-100MG
4-[(tert-Butoxycarbonylamino)methyl]pyridine-2-carboxylic acid ≥96.0% (by HPLC, titration analysis)
4-[(tert-Butoxycarbonylamino)methyl]pyridine-2-carboxylic acid

[Reagent for application of the exciton chirality method]
CAS Number: 260970-57-8
MDL Number: MFCD02093452
Molecular Formula: C12H16N2O4
Molecular Weight: 252.27
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal


Product Categories: Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: 4-(Boc-aminomethyl)pyridine-2-carboxylic Acid

Formula: C₁₂H₁₆N₂O₄
MW: 252.27 g/mol
MDL Number: MFCD02093452
CAS Number: 260970-57-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eed45a7bb1905036e8242f4c4f946be0fe5c199a&vid=eed45a7bb1905036e8242f4c4f946be0fe5c199a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR