(S)-1-Boc-2-azetidinemethanol ≥95.0%
Supplier: TCI America
B2123-100MG
B2123-1G
TCB2123-100MGEA
390.1
CAD
TCB2123-100MG
TCB2123-1G
(S)-1-Boc-2-azetidinemethanol ≥95.0%
(S)-1-Boc-2-azetidinemethanol
CAS Number: 161511-85-9
MDL Number: MFCD03093620
Molecular Formula: C9H17NO3
Molecular Weight: 187.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.05
Specific rotation [a]20/D: -74 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-1-Boc-2-azetidinemethanol
Formula:
C₉H₁₇NO₃ Density: 1.05 (20 °C) |
MDL Number:
MFCD03093620 CAS Number: 161511-85-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...