3,5-Bis[3,5-bis(benzyloxy)benzyloxy]benzyl bromide ≥97.0% (by HPLC)
Supplier: TCI America
B2118-1G
B2118-5G
TCB2118-1GEA
360.24
CAD
TCB2118-1G
TCB2118-5G
3,5-Bis[3,5-bis(benzyloxy)benzyloxy]benzyl bromide ≥97.0% (by HPLC)
3,5-Bis[3,5-bis(benzyloxy)benzyloxy]benzyl bromide
CAS Number: 129536-41-0
MDL Number: MFCD02093449
Molecular Formula: C49H43BrO6
Molecular Weight: 807.78
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 129
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 3,5-Bis(3,5-dibenzyloxybenzyloxy)benzyl Bromide
Formula:
C₄₉H₄₃BrO₆ Melting Pt: 129 °C |
MDL Number:
MFCD02093449 CAS Number: 129536-41-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...