3-(2-Benzimidazolyl)-7-(diethylamino)coumarin ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2111-1G B2111-100MG
TCB2111-001GEA 317.58 CAD
TCB2111-001G TCB2111-100MG
3-(2-Benzimidazolyl)-7-(diethylamino)coumarin ≥98.0% (by titrimetric analysis)
3-(2-Benzimidazolyl)-7-(diethylamino)coumarin

CAS Number: 27425-55-4
MDL Number: MFCD00051348
Molecular Formula: C20H19N3O2
Molecular Weight: 333.39
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Coumarin Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: Coumarin 7

Formula: C₂₀H₁₉N₃O₂
MDL Number: MFCD00051348
CAS Number: 27425-55-4
EINECS: 248-451-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=821282a13ab4db50a2a019491eb5249ff00e8148&vid=821282a13ab4db50a2a019491eb5249ff00e8148&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR