1,3-Bis(benzyloxy)-2-propanol ≥96.0%

Supplier: TCI America

Synonyms: 1,3-Dibenzyloxy-2-propanol, 1,3-Di-O-benzylglycerol

B2110-25G B2110-5G
TCB2110-25GEA 479.1 CAD
TCB2110-25G TCB2110-5G
1,3-Bis(benzyloxy)-2-propanol ≥96.0%
1,3-Bis(benzyloxy)-2-propanol

CAS Number: 6972-79-8
MDL Number: MFCD00010616
Molecular Formula: C17H20O3
Molecular Weight: 272.34
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.10
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1,3-Di-O-benzylglycerol, 1,3-Dibenzyloxy-2-propanol

Formula: C₁₇H₂₀O₃
MW: 272.34 g/mol
Density: 1.10 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00010616
CAS Number: 6972-79-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1967da882496a47043ae410141baffd4dde6bc1&vid=c1967da882496a47043ae410141baffd4dde6bc1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR