Benzyl glycidyl ether ≥97.0%
Supplier: TCI America
Synonyms:
2-((Benzyloxy)methyl)oxirane
B2108-25G
B2108-5G
TCB2108-25GEA
159.58
CAD
TCB2108-25G
TCB2108-5G
Benzyl glycidyl ether ≥97.0%
Benzyl glycidyl ether
CAS Number: 2930-05-4
MDL Number: MFCD00068664
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 75
Flash Point (°C): 110
Specific Gravity (20/20): 1.07
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents
Other Product Names / Synonyms: 1-Benzyloxy-2,3-epoxypropane, 2-(Benzyloxymethyl)oxirane
Formula:
C₁₀H₁₂O₂ MW: 164.2011 g/mol Boiling Pt: 75 °C Density: 1.07 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00068664 CAS Number: 2930-05-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...