(1S,2R)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol ≥98.0%

Supplier: TCI America

Synonyms: (1S,2R)-N-Benzyl-N-(mesitylenesulfonyl)norephedrine, (1S,2R)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulfonyl)amino]-1-phenyl-1-propanol, (1S,2R)-N-Benzyl-N-(mesitylenesulphonyl)norephedrine, (1S,2R)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulphonyl)amino]-1-phenyl-1-propanol

B2104-5G B2104-1G
TCB2104-5GEA 736.28 CAD
TCB2104-5G TCB2104-001G
(1S,2R)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol ≥98.0%
(1S,2R)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol

CAS Number: 187324-64-7
MDL Number: MFCD02093447
Molecular Formula: C25H29NO3S
Molecular Weight: 423.57
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -49 deg (C=2, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Auxiliaries
Other Product Names / Synonyms: (1S,2R)-N-Benzyl-N-(mesitylenesulfonyl)norephedrine, (1S,2R)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulfonyl)amino]-1-phenyl-1-propanol

Formula: C₂₅H₂₉NO₃S
MDL Number: MFCD02093447
CAS Number: 187324-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7556a4da3c7ff976e4144ac6d29b0139f3fa3315&vid=7556a4da3c7ff976e4144ac6d29b0139f3fa3315&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR