(1R,2S)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol ≥98.0%

Supplier: TCI America

Synonyms: (1R,2S)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulfonyl)amino]-1-phenyl-1-propanol, (1R,2S)-N-Benzyl-N-(mesitylenesulfonyl)norephedrine, (1R,2S)-N-Benzyl-N-(mesitylenesulphonyl)norephedrine, (1R,2S)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulphonyl)amino]-1-phenyl-1-propanol

B2103-1G B2103-5G
TCB2103-001GEA 203.35 CAD
TCB2103-001G TCB2103-5G
(1R,2S)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol ≥98.0%
(1R,2S)-2-[N-Benzyl-N-(mesitylenesulfonyl)amino]-1-phenyl-1-propanol

CAS Number: 187324-63-6
MDL Number: MFCD02093446
Molecular Formula: C25H29NO3S
Molecular Weight: 423.57
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 132
Specific rotation [a]20/D: 50 deg (C=2, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Auxiliaries
Other Product Names / Synonyms: (1R,2S)-N-Benzyl-N-(mesitylenesulfonyl)norephedrine, (1R,2S)-2-[N-Benzyl-N-(2,4,6-trimethylbenzenesulfonyl)amino]-1-phenyl-1-propanol

Formula: C₂₅H₂₉NO₃S
Melting Pt: 132 °C
MDL Number: MFCD02093446
CAS Number: 187324-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89a8912ccb91b428e32fd3d210ef0d9367860b1d&vid=89a8912ccb91b428e32fd3d210ef0d9367860b1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR