2-(4-Bromophenyl)-1,3-dioxolane ≥98.0%
Supplier: TCI America
Synonyms:
4-Bromobenzaldehyde Ethylene Acetal
B2100-25G
B2100-5G
TCB2100-25GEA
477.16
CAD
TCB2100-25G
TCB2100-005G
2-(4-Bromophenyl)-1,3-dioxolane ≥98.0%
2-(4-Bromophenyl)-1,3-dioxolane
CAS Number: 10602-01-4
MDL Number: MFCD01318950
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 35
Other Product Names / Synonyms: 4-Bromobenzaldehyde Ethylene Acetal
Formula:
C₉H₉BrO₂ |
MDL Number:
MFCD01318950 CAS Number: 10602-01-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...