Trihexyl borate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Boric Acid Trihexyl Ester

B2094-25G B2094-5G
TCB2094-25GEA 192.76 CAD
TCB2094-25G TCB2094-5G
Trihexyl borate ≥98.0% (by titrimetric analysis)
Trihexyl borate

CAS Number: 5337-36-0
MDL Number: MFCD00027295
Molecular Formula: C18H39BO3
Molecular Weight: 314.32
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 311
Flash Point (°C): 149
Specific Gravity (20/20): 0.85


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Boric Acid Trihexyl Ester

Formula: C₁₈H₃₉BO₃
MW: 314.32 g/mol
Boiling Pt: 311 °C
Density: 0.85 (20 °C)
Flash Pt: 149 °C
MDL Number: MFCD00027295
CAS Number: 5337-36-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48f6548378179caab0fdc7d03f8667a1f010d222&vid=48f6548378179caab0fdc7d03f8667a1f010d222&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR