Methyl-3,5-dibenzyloxybenzoate ≥98.0%

Supplier: TCI America

B2092-5G B2092-25G
TCB2092-005GEA 47.4 CAD
TCB2092-005G TCB2092-25G
Methyl-3,5-dibenzyloxybenzoate ≥98.0%
Methyl-3,5-dibenzyloxybenzoate

CAS Number: 58605-10-0
MDL Number: MFCD00157205
Molecular Formula: C22H20O4
Molecular Weight: 348.40
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 71


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: 3,5-Bis(benzyloxy)benzoic Acid Methyl Ester, 3,5-Dibenzyloxybenzoic Acid Methyl Ester

Formula: C₂₂H₂₀O₄
Melting Pt: 71 °C
MDL Number: MFCD00157205
CAS Number: 58605-10-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bdcd198198f4e4d9c85120a09b54537a5fff7e4&vid=11272d7e365359391050d8c502e2c24f2fe3028b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR