2-Bromoethyl phenyl sulfide ≥97.0%

Supplier: TCI America

B2085-25G B2085-5G
TCB2085-25GEA 361.82 CAD
TCB2085-25G TCB2085-005G
2-Bromoethyl phenyl sulfide ≥97.0%
2-Bromoethyl phenyl sulphide

CAS Number: 4837-01-8
MDL Number: MFCD00040826
Molecular Formula: C8H9BrS
Molecular Weight: 217.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Specific Gravity (20/20): 1.44
Storage Temperature: 0-10°C

Formula: C₈H₉BrS
Boiling Pt: 137 °C
Density: 1.44 (20 °C)
MDL Number: MFCD00040826
CAS Number: 4837-01-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02a981dca38aeabfc68f734487434c3b233bfbbe&vid=02a981dca38aeabfc68f734487434c3b233bfbbe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR