Diethyl (5-bromopentyl)malonate ≥97.0%
Supplier: TCI America
B2083-25G
B2083-5G
TCB2083-025GEA
270.18
CAD
TCB2083-025G
TCB2083-005G
Diethyl (5-bromopentyl)malonate ≥97.0%
Diethyl (5-bromopentyl)malonate
CAS Number: 1906-95-2
MDL Number: MFCD00048565
Molecular Formula: C12H21BrO4
Molecular Weight: 309.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 161
Specific Gravity (20/20): 1.23
Other Product Names / Synonyms: (5-Bromopentyl)malonic Acid Diethyl Ester
Formula:
C₁₂H₂₁BrO₄ Boiling Pt: 161 °C Density: 1.23 (20 °C) |
MDL Number:
MFCD00048565 CAS Number: 1906-95-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...