3,5-Bis(4-aminophenoxy)benzoic acid ≥96.0% (by HPLC)

Supplier: TCI America

B2081-1G B2081-5G
TCB2081-1GEA 244.58 CAD
TCB2081-1G TCB2081-005G
3,5-Bis(4-aminophenoxy)benzoic acid ≥96.0% (by HPLC)
3,5-Bis(4-aminophenoxy)benzoic acid

CAS Number: 195189-45-8
MDL Number: MFCD02093443
Molecular Formula: C19H16N2O4
Molecular Weight: 336.35
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Melting point (°C): 240

Formula: C₁₉H₁₆N₂O₄
Melting Pt: 240 °C
MDL Number: MFCD02093443
CAS Number: 195189-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f25661403f9a0bfd98f76cda7427a85fae9ae679&vid=f25661403f9a0bfd98f76cda7427a85fae9ae679&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR