1,1-Bis[4-[N,N-di(p-tolyl)amino]phenyl]cyclohexane ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
TAPC, 4,4'-Cyclohexylidenebis[N,N-di(p-tolyl)aniline]
B2079-1G
B2079-5G
TCB2079-1GEA
159.58
CAD
TCB2079-1G
TCB2079-5G
1,1-Bis[4-[N,N-di(p-tolyl)amino]phenyl]cyclohexane ≥98.0% (by total nitrogen basis)
1,1-Bis[4-[N,N-di(p-tolyl)amino]phenyl]cyclohexane
CAS Number: 58473-78-2
MDL Number: MFCD03844768
Molecular Formula: C46H46N2
Molecular Weight: 626.89
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 186
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials
Other Product Names / Synonyms: 4,4'-Cyclohexylidenebis[N,N-di(p-tolyl)aniline], TAPC
Formula:
C₄₆H₄₆N₂ MW: 626.89 g/mol Melting Pt: 186 °C |
MDL Number:
MFCD03844768 CAS Number: 58473-78-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...