Bis[2-(2-benzothiazolyl)phenolato]zinc(II) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2077-5G B2077-1G
TCB2077-5GEA 150.1 CAD
TCB2077-5G TCB2077-1G
Bis[2-(2-benzothiazolyl)phenolato]zinc(II) ≥98.0% (by titrimetric analysis)
Bis[2-(2-benzothiazolyl)phenolato]zinc(II)

CAS Number: 58280-31-2
MDL Number: MFCD03844766
Molecular Formula: C26H16N2O2S2Zn
Molecular Weight: 517.93
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₂₆H₁₆N₂O₂S₂Zn
MDL Number: MFCD03844766
CAS Number: 58280-31-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c135ea9bb632592354f1cfba1b2a5a85c1e4af44&vid=c135ea9bb632592354f1cfba1b2a5a85c1e4af44&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR