1,1-Bis(4-aminophenyl)cyclohexane ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Cyclohexylidenedianiline
B2076-5G
B2076-25G
TCB2076-5GEA
338.91
CAD
TCB2076-5G
TCB2076-25G
1,1-Bis(4-aminophenyl)cyclohexane ≥98.0%
1,1-Bis(4-aminophenyl)cyclohexane
CAS Number: 3282-99-3
MDL Number: MFCD00035700
Molecular Formula: C18H22N2
Molecular Weight: 266.39
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 110
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 4,4'-Cyclohexylidenedianiline
Formula:
C₁₈H₂₂N₂ MW: 266.39 g/mol Melting Pt: 110 °C |
MDL Number:
MFCD00035700 CAS Number: 3282-99-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...